Learn More
MSC Ergodyne Hi-Vis Safety Vest, Class 2, Hook-and-Loop, S/M, 1 ea
Catalog No. 5000766807
Encompass
The GloWear 8205HL Hi-Vis Safety Vest is designed to enhance visibility in low-light conditions, providing ANSI/ISEA 107-2020 Type R, Class 2 compliance. Constructed from 3.3oz polyester mesh fabric, it offers all-day comfort and a secure fit with a hook and loop closure.
  • Increased visibility with 2-inch reflective, solid tape
  • All-day comfort with polyester mesh fabric (3.3oz)
  • Easy on and off with hook and loop closure
  • Easy maintenance with machine-washable fabric, retaining shape for up to 25 washes
Catalog No. 50-007-66807 Supplier MSC Supplier No. 32762221
The GloWear 8205HL Hi-Vis Safety Vest is designed to enhance visibility in low-light conditions, providing ANSI/ISEA 107-2020 Type R, Class 2 compliance. Constructed from 3.3oz polyester mesh fabric, it offers all-day comfort and a secure fit with a hook and loop closure.
  • Increased visibility with 2-inch reflective, solid tape
  • All-day comfort with polyester mesh fabric (3.3oz)
  • Easy on and off with hook and loop closure
  • Easy maintenance with machine-washable fabric, retaining shape for up to 25 washes