Learn More
Enterprise Technology Solutions Ergodyne Holder,f/PPE Gear,Dual Clips,Breakaway,6-1/2"x1"x1",Lime
SDP
Catalog No. 5000831261
Encompass_Preferred
Squids 3400 Glove Clip Holder with dual clips keeps your face masks, gloves, ice traction, cooling towels, keys and whatever else always seems to mysteriously disappear close at hand. Versatile dual clip can be used as one unit or detached and used as two separate pieces. Breakaway feature allows the glove grabbers to safely break away in an emergency. Patented design allows it to be put back together and used again. Strong gripping jaws can hold onto gloves, hats, keys, towels and other PPE that are too bulky to fit in pockets. Acetal copolymer construction is nonconductive, temperature-resistant and chemical-resistant. Opening of the grabber's teeth is 1/2". Average breaking strength is 26 lb.
Catalog No. 50-008-31261 Supplier Enterprise Technology Solutions Supplier No. EGO19119
Squids 3400 Glove Clip Holder with dual clips keeps your face masks, gloves, ice traction, cooling towels, keys and whatever else always seems to mysteriously disappear close at hand. Versatile dual clip can be used as one unit or detached and used as two separate pieces. Breakaway feature allows the glove grabbers to safely break away in an emergency. Patented design allows it to be put back together and used again. Strong gripping jaws can hold onto gloves, hats, keys, towels and other PPE that are too bulky to fit in pockets. Acetal copolymer construction is nonconductive, temperature-resistant and chemical-resistant. Opening of the grabber's teeth is 1/2". Average breaking strength is 26 lb.