Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 325 Non-Marring Rubber Cap Knee Pads

Catalog No. 19046106 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Non-Marring Knee Pad
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-046-106 Non-Marring Knee Pad
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-046-106 Supplier Ergodyne™ Supplier No. 18325
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Color Black, Tan
Closure Type Buckle
Product Type Proflex™ Long Cap Knee pad
Design Nickel-plated grommets, anti-odor treatment, contoured side cut
Size Universal
For Use With (Application) Construction, masonry, freight or baggage, flooring, maintenance, utilities, carpentry
Liner Material 0.5 in. EVA
Model 320
Strap Material Woven Elastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.