Learn More
Ergodyne™ ProFlex™ 4020 Wrist Supports
Catalog No. 19035280E Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
Size:
Large/X-Large
Medium
X-Small/Small
Hand:
Left
Right
Model:
4020
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Hand Model
19-035280E Blue Large/X-Large Left 4020
19-035280D Blue Medium Left 4020
19-035279D Blue Medium Right 4020
19-035279C Blue X-Small/Small Right 4020
19-035280C Blue X-Small/Small Left 4020
19-035279E Blue Large/X-Large Right 4020
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-035280E Supplier Ergodyne™ Supplier No. 70246
Limit downward wrist movement during repetitive work
  • Lightweight, compact design helps to maintain a neutral position
  • Patented Open-Center Stay™
  • Contoured design follows natural shape of wrist for exact fit
  • Neoprene body cloth conforms to wrist and hands for comfortable, glovelike fit
  • Interior ventilation holes keep hands dry for added comfort
  • Machine washable
  • Blue
Order Info
Sold individually
Specifications
Color Blue
Closure Type Two stage hook and loop
Product Type Proflex™ Wrist Support
Design Contoured
Size Large/X-Large
For Use With (Application) Designed to relieve stress associated with repetitive strain and overuse injuries and also for food processing, picking, packing, assembly, fabrication, office, retail and light industrial
Includes Patented open-center Stay™, interior ventilation holes
Hand Left
Limitations Limit downward wrist movement
Material Neoprene
Model 4020
Size (English) 7 to 8 in.
Size (Metric) 18 to 20 cm