ProFlex 810 Reinforced Utility Gloves are designed for long-lasting performance and all-day comfort in demanding jobs. Featuring a durable AX Suede palm reinforcement, these gloves deliver a tougher grip and longer life, standing up to repeated use and heavy handling without wearing thin.
- Ultra-durable AX Suede reinforcements on the palm and thumb saddle strengthen high-stress areas for secure grip and extended durability.
- Poly mesh construction promotes airflow to keep hands cool and comfortable.
- Molded hook and loop closure with ID space stays out of the way while keeping gloves snug.
- Integrated pull tab simplifies wear and removal between tasks.
- Meets EN 388: 3121X standard for mechanical protection.
- Ideal for material handling, construction, maintenance, and light-to-medium duty applications.