Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 9002 Certified Anti-Vibration Gloves

Catalog No. 19050509 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-050-509 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-050-509 Supplier Ergodyne™ Supplier No. 17322

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed of pigskin leather for durability, while a Nu2O2™ polymer pad provides a high level of anti-vibration protection.

  • Pigskin leather fingers and palm
  • Neoprene knuckle pad
  • Woven elastic cuff
  • Nu2O2™ polymer pad
  • Built in wrist support with patented Open-Center Stay™ technology

Specifications

Certifications/Compliance ANSI S2.73, ISO 10819:1996
Cuff Material Elastic
Color Black
Hand Specific
Material Pigskin Leather
Product Type Anti-vibration Gloves
Size Small
For Use With (Application) Demolition, Heavy Construction, and Oil/Gas Drilling
Series ProFlex™ 9002
Length (English) 7 in.
Length (Metric) 17.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.