Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 9012 Certified Anti-Vibration Gloves with Wrist Support

Catalog No. 191500114A Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-0114A Small
19-150-0114B Medium
19-150-0114C Large
19-150-0114D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-150-0114A Supplier Ergodyne™ Supplier No. 17722
This item is not returnable. View return policy

Gloves feature a patented Nu202™ polymer pad on the palm for additional protection, and built-in wrist support with Open-Center Stay™ technology.

  • Patented Nu2O2™ polymer pad
  • Pigskin leather palm and fingers
  • Built-in wrist support with patented Open-Center Stay™ technology
  • Low-profile closure with woven elastic cuff
  • Neoprene knuckle pad
  • Anti-odor treatment

Specifications

Certifications/Compliance ANSI S2.73/ISO 10819:1996
Cuff Material Elastic
Color Black
Hand Specific
Material Pigskin Leather
Product Type Anti-vibration Gloves
Size Small
For Use With (Application) Power Tools, Repetitive Motion
Glove Cuff Style Hook and Loop
Series ProFlex™ 9012
Length (English) 7 in.
Length (Metric) 17.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.