Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 9015F(x) Certified Anti-Vibration Gloves with Dorsal Protection

Catalog No. 19052625D Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-052-625D 2X-Large
19-052-625A Medium
19-052-625B Large
19-052-625C X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-052-625D Supplier Ergodyne™ Supplier No. 16236

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pigskin leather gloves are constructed with molded TPR to offer enhanced impact protection, while a patented Nu2O2™ polymer pad minimizes vibration.

  • Maximum shock, impact, and vibration protection with exclusive, patented Nu2O2™ polymer
  • Pigskin leather palm and fingers for rugged wear and abrasion resistance
  • PVC shell for extra protection for the back of the hand
  • Finger tip reinforcement
  • Micro-injected closure for quick cinching
  • Woven elastic cuff for a perfect fit
  • Wicked™ treated
  • Pull-on tab

Specifications

Certifications/Compliance ANSI S3.40, ISO 10819-1996
Cuff Material Woven Elastic
Color Black
Hand Specific
Material Pigskin Leather
Product Type Anti-vibration Gloves
Size 2X-Large
For Use With (Application) Demolition, Heavy Construction, Mining, Oil and Gas Refining
Glove Cuff Style Open
Series Proflex™ 9015F(x)
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.