Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 400 Universal Wrist Wrap

Catalog No. 19037825 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Tan
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-037-825 Black
19-037-826 Tan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-037-825 Supplier Ergodyne™ Supplier No. 72102
Add to Cart
Add to Cart

Provide wrist support for a variety of applications

  • Durable woven elastic
  • Single hook and loop closure
  • Ambidextrous; one size

Food processing, Picking/packing, Assembly/fabrication

Specifications

Color Black
Closure Type Hook and loop
Product Type Proflex™ Wrist Wrap
Size Universal
For Use With (Application) Food processing, picking, packing, assembly, fabrication
Hand Ambidextrous
Material Woven Elastic
Model 400
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.