Learn More
Enterprise Technology Solutions Ergodyne Safety Glasses,142mm W,Anti-fog CL Lens/Black Frame
SDP
Catalog No. 5000828025
Encompass_Preferred
Skullerz Odin Safety Glasses feature a tough, scratch-resistant lens that filters 99.9 percent of harmful UVA, UVB and UVC rays to keep your eyes safe in the sun. They are military tested and ANSI Compliant. Clear Fog-off Lens is ideal for indoor use, low light and hazy/cloudy conditions. Permanent, anti-fog treatment delivers fog-free viewing at all times and will not wash or wear off. Great for use with dust masks, these lenses won't fog so you can focus on the job at hand. Durable, nylon frames bend and flex without breaking. Nonslip, rubber nose piece and rubber temples keep the full-frame glasses secure and resist sweat. Integrated accessory receptors at each temple also improve airflow when not in use. Frame width is 142mm.
Catalog No. 50-008-28025 Supplier Enterprise Technology Solutions Supplier No. EGO50003
Skullerz Odin Safety Glasses feature a tough, scratch-resistant lens that filters 99.9 percent of harmful UVA, UVB and UVC rays to keep your eyes safe in the sun. They are military tested and ANSI Compliant. Clear Fog-off Lens is ideal for indoor use, low light and hazy/cloudy conditions. Permanent, anti-fog treatment delivers fog-free viewing at all times and will not wash or wear off. Great for use with dust masks, these lenses won't fog so you can focus on the job at hand. Durable, nylon frames bend and flex without breaking. Nonslip, rubber nose piece and rubber temples keep the full-frame glasses secure and resist sweat. Integrated accessory receptors at each temple also improve airflow when not in use. Frame width is 142mm.