Learn More
Enterprise Technology Solutions Ergodyne Safety Glasses,142mm W,Smoke Lens/Black Frame
SDP
Catalog No. 5000828213
Encompass_Preferred
Skullerz Odin Safety Glasses feature a tough, scratch-resistant lens that filters 99.9 percent of harmful UVA, UVB and UVC rays to keep your eyes safe in the sun. They are military tested and ANSI Compliant. This smoke lens is ideal for outdoors and bright light. Durable, nylon frames bend and flex without breaking. Nonslip, rubber nose piece and rubber temples keep the full-frame glasses secure and resist sweat. Integrated accessory receptors at each temple also improve airflow when not in use. Frame width is 142mm.
Catalog No. 50-008-28213 Supplier Enterprise Technology Solutions Supplier No. EGO50030
Skullerz Odin Safety Glasses feature a tough, scratch-resistant lens that filters 99.9 percent of harmful UVA, UVB and UVC rays to keep your eyes safe in the sun. They are military tested and ANSI Compliant. This smoke lens is ideal for outdoors and bright light. Durable, nylon frames bend and flex without breaking. Nonslip, rubber nose piece and rubber temples keep the full-frame glasses secure and resist sweat. Integrated accessory receptors at each temple also improve airflow when not in use. Frame width is 142mm.