Learn More
MSC Ergodyne Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, 1 ea
Catalog No. 5000687105
Encompass
These safety glasses offer reliable eye protection for indoor and outdoor use, featuring anti-fog and scratch-resistant properties to ensure clear visibility in various conditions. The frameless design and wrap-around style provide a universal fit and comprehensive coverage.
  • Anti-fog and scratch-resistant polycarbonate lenses
  • Frameless, wrap-around frame design
  • Clear mirror lens color
  • Provides UV protection
  • Impact-resistant
  • Meets CSA Z94.3, EN 166, and ANSI Z87.1+ safety standards
  • Flexible nylon frame temples for comfort
Catalog No. 50-006-87105 Supplier MSC Supplier No. 28762169
These safety glasses offer reliable eye protection for indoor and outdoor use, featuring anti-fog and scratch-resistant properties to ensure clear visibility in various conditions. The frameless design and wrap-around style provide a universal fit and comprehensive coverage.
  • Anti-fog and scratch-resistant polycarbonate lenses
  • Frameless, wrap-around frame design
  • Clear mirror lens color
  • Provides UV protection
  • Impact-resistant
  • Meets CSA Z94.3, EN 166, and ANSI Z87.1+ safety standards
  • Flexible nylon frame temples for comfort