Learn More
Enterprise Technology Solutions Ergodyne Safety Glasses,Blade Style,Anti-Fog,Clear Lens/BK
SDP
Catalog No. 5000828000
Encompass_Preferred
Aegir Anti-Scratch & Enhanced Anti-Fog Safety Glasses feature an oversized, blade-style design, providing superior eye protection and maximized field of vision. Tough, scratch-resistant, anti-fog, polycarbonate lenses protect against impacts and filter 99.9 percent of harmful UVA, UVB and UVC rays. High-strength frame is built for durability. Rubberized nose bridge and temples offer nonslip security and resist sweat for all-day comfort. Rubber temples bend and re-bend to accommodate a variety of head shapes and fit preferences. Fog-Off Plus technology combines durable scratch-resistance with enhanced, anti-fog performance for clear viewing no matter the job. Impact-resistant safety glasses are tested to ANSI/ISEA Z87.1-2020 High-Velocity Impact Protection and optical requirements. They are perfect for outdoor work such as construction, landscaping, carpentry, woodworking and more. Clear lenses with 90 percent VLT are ideal for indoor, low-light and nighttime conditions.
Catalog No. 50-008-28000 Supplier Enterprise Technology Solutions Supplier No. EGO55002
Aegir Anti-Scratch & Enhanced Anti-Fog Safety Glasses feature an oversized, blade-style design, providing superior eye protection and maximized field of vision. Tough, scratch-resistant, anti-fog, polycarbonate lenses protect against impacts and filter 99.9 percent of harmful UVA, UVB and UVC rays. High-strength frame is built for durability. Rubberized nose bridge and temples offer nonslip security and resist sweat for all-day comfort. Rubber temples bend and re-bend to accommodate a variety of head shapes and fit preferences. Fog-Off Plus technology combines durable scratch-resistance with enhanced, anti-fog performance for clear viewing no matter the job. Impact-resistant safety glasses are tested to ANSI/ISEA Z87.1-2020 High-Velocity Impact Protection and optical requirements. They are perfect for outdoor work such as construction, landscaping, carpentry, woodworking and more. Clear lenses with 90 percent VLT are ideal for indoor, low-light and nighttime conditions.