Learn More
Enterprise Technology Solutions Ergodyne Safety Glasses,Blade Style,Smoke Lens/BK Frame
SDP
Supplier:  Enterprise Technology Solutions EGO55005
Encompass_Preferred
Aegir Safety Glasses feature an oversized, blade-style design, providing superior eye protection and maximized field of vision. Polycarbonate lenses protect against impacts and filter 99.9 percent of harmful UVA, UVB and UVC rays. High-strength frame is built for durability. Rubberized nose bridge and temples offer nonslip security and resist sweat for all-day comfort. Rubber temples bend and re-bend to accommodate a variety of head shapes and fit preferences. Impact-resistant safety glasses are tested to ANSI/ISEA Z87.1-2020 High-Velocity Impact Protection and optical requirements. They are perfect for outdoor work such as construction, landscaping, carpentry, woodworking and more. Smoke lenses with 20 percent VLT are ideal for outdoor and bright-light conditions.
Catalog No. 50-008-28027