Learn More
MSC Ergodyne Safety Glasses, Clear Lens, Black Frame, Z87.1-2020, 1 ea
Catalog No. 5000677965
Encompass
The Skullerz DAGR Safety Glasses offer on-the-job protection and off-duty style with a sleek, blade-style, half-frame design. Durable nylon frames bend and flex without breaking. Polycarbonate lenses provide robust impact protection and filter 99.9% of harmful UVA, UVB, and UVC rays. A non-slip rubber nose piece and temples ensure a secure fit.
  • Impact resistant: meets ANSI/ISEA Z87.1-2020 (Mark: Z87.1+), CSA Z94.3, and MIL-PRF 32432 Ballistic Impact standards.
  • Durable frame: flexible nylon frame ensures extended product life and resilience.
  • Half-frame style: improves lower and peripheral vision due to its frameless bottom.
  • UV protection: lenses filter 99.9% of harmful UVA, UVB, and UVC rays.
  • Non-slip design: rubber nose piece and temples keep glasses securely in place.
Catalog No. 50-006-77965 Supplier MSC Supplier No. 31782378
The Skullerz DAGR Safety Glasses offer on-the-job protection and off-duty style with a sleek, blade-style, half-frame design. Durable nylon frames bend and flex without breaking. Polycarbonate lenses provide robust impact protection and filter 99.9% of harmful UVA, UVB, and UVC rays. A non-slip rubber nose piece and temples ensure a secure fit.
  • Impact resistant: meets ANSI/ISEA Z87.1-2020 (Mark: Z87.1+), CSA Z94.3, and MIL-PRF 32432 Ballistic Impact standards.
  • Durable frame: flexible nylon frame ensures extended product life and resilience.
  • Half-frame style: improves lower and peripheral vision due to its frameless bottom.
  • UV protection: lenses filter 99.9% of harmful UVA, UVB, and UVC rays.
  • Non-slip design: rubber nose piece and temples keep glasses securely in place.