Learn More
MSC Ergodyne Safety Glasses, Polycarbonate, Smoke Lens, Scratch-Resistant, 1 ea
Catalog No. 5000795785
Encompass
These safety glasses feature scratch-resistant polycarbonate smoke lenses with a half-framed design, suitable for environments requiring robust eye protection. They offer a black frame with non-adjustable temples and meet ANSI Z87.1+, CSA Z94.3, and MIL-PRF 32432 ballistic impact standards.
  • Tinted glasses style
  • Scratch-resistant lens coating
  • Half-framed style with wrap-around design
  • Smoke lens color
  • Black frame and temple color
  • Universal size
  • Polycarbonate lens material
  • Non-adjustable temples
  • UV protection
  • Meets CSA Z94.3, MIL-PRF 32432 Ballistic Impact, and ANSI Z87.1+ standards
  • Impact-resistant
  • Rubber frame material
Catalog No. 50-007-95785 Supplier MSC Supplier No. 31782428
These safety glasses feature scratch-resistant polycarbonate smoke lenses with a half-framed design, suitable for environments requiring robust eye protection. They offer a black frame with non-adjustable temples and meet ANSI Z87.1+, CSA Z94.3, and MIL-PRF 32432 ballistic impact standards.
  • Tinted glasses style
  • Scratch-resistant lens coating
  • Half-framed style with wrap-around design
  • Smoke lens color
  • Black frame and temple color
  • Universal size
  • Polycarbonate lens material
  • Non-adjustable temples
  • UV protection
  • Meets CSA Z94.3, MIL-PRF 32432 Ballistic Impact, and ANSI Z87.1+ standards
  • Impact-resistant
  • Rubber frame material