Learn More
MSC Ergodyne Skullerz Glasses, Yellow, Black, ANSI Z87.1, 1 ea
Catalog No. 5000675965
Encompass
Skullerz DAGR Safety Glasses with yellow lenses provide superior eye protection with a sleek, sporty, half-frame design. These glasses feature polycarbonate lenses that protect against impacts and filter 99.9% of harmful UVA, UVB, and UVC rays. Their durable nylon frame is designed to flex without breaking, complemented by a non-slip rubber nose piece and temples for a secure fit, making them suitable for indoor, overcast, flat, and low light conditions.
  • Meets ANSI/ISEA Z87.1-2020 and MIL-PRF 32432 Ballistic Impact standards
  • Durable nylon blade-style frame bends and flexes without breaking
  • Half-frame style improves lower and peripheral vision
  • Lenses filter 99.9% of harmful UVA, UVB, and UVC rays
  • Non-slip rubber nose piece and temples resist sweat
  • Yellow lenses offer 91% visual light transmission for low light conditions
Catalog No. 50-006-75965 Supplier MSC Supplier No. 31782402
Skullerz DAGR Safety Glasses with yellow lenses provide superior eye protection with a sleek, sporty, half-frame design. These glasses feature polycarbonate lenses that protect against impacts and filter 99.9% of harmful UVA, UVB, and UVC rays. Their durable nylon frame is designed to flex without breaking, complemented by a non-slip rubber nose piece and temples for a secure fit, making them suitable for indoor, overcast, flat, and low light conditions.
  • Meets ANSI/ISEA Z87.1-2020 and MIL-PRF 32432 Ballistic Impact standards
  • Durable nylon blade-style frame bends and flexes without breaking
  • Half-frame style improves lower and peripheral vision
  • Lenses filter 99.9% of harmful UVA, UVB, and UVC rays
  • Non-slip rubber nose piece and temples resist sweat
  • Yellow lenses offer 91% visual light transmission for low light conditions