Learn More
MSC Ergodyne Skullerz THOR Safety Glasses, Polycarbonate, Anti-Fog, 1 ea
Catalog No. 5000686272
Encompass
Ergodyne Skullerz THOR Safety Glasses are designed for robust eye protection in demanding environments. They feature anti-fog and scratch-resistant polycarbonate lenses for clear vision and durability.
  • Polycarbonate lens material
  • Blue mirror lens color
  • Full-framed, wrap-around design
  • Anti-fog and scratch-resistant coatings
  • Provides UV protection
  • Impact-resistant
  • Meets ANSI Z87.1+, CSA Z94.3, and MIL-PRF 32432 Ballistic Impact standards
Catalog No. 50-006-86272 Supplier MSC Supplier No. 31782121
Ergodyne Skullerz THOR Safety Glasses are designed for robust eye protection in demanding environments. They feature anti-fog and scratch-resistant polycarbonate lenses for clear vision and durability.
  • Polycarbonate lens material
  • Blue mirror lens color
  • Full-framed, wrap-around design
  • Anti-fog and scratch-resistant coatings
  • Provides UV protection
  • Impact-resistant
  • Meets ANSI Z87.1+, CSA Z94.3, and MIL-PRF 32432 Ballistic Impact standards