Learn More
MSC Ergodyne Strap-On Cleat, Stud Traction, 8 to 11, 1 ea
Catalog No. 5000686977
Encompass
This strap-on cleat is designed for enhanced stud traction, offering a secure and adjustable fit for various work boots.
  • Features stud traction
  • Adjustable cord attachment
  • Compatible with men's shoe sizes 8 to 11
  • Fits most work boots
  • Black in color
  • Replacement spike compatibility TREX 6301
Catalog No. 50-006-86977 Supplier MSC Supplier No. 40972291
This strap-on cleat is designed for enhanced stud traction, offering a secure and adjustable fit for various work boots.
  • Features stud traction
  • Adjustable cord attachment
  • Compatible with men's shoe sizes 8 to 11
  • Fits most work boots
  • Black in color
  • Replacement spike compatibility TREX 6301