Learn More
Enterprise Technology Solutions Ergodyne Sunglasses/Goggles,Indoor/Outdoor Lens/Black Frame
SDP
Catalog No. 5000828417
Encompass_Preferred
Skullerz Loki Safety Glasses are designed to be worn as regular sunglasses or as safety goggles by removing the temples and attaching the elastic strap and removable foam gasket for a dust-beating seal (both included). Tough, scratch-resistant lens filters 99.9 percent of harmful UVA, UVB and UVC rays to keep your eyes safe in the sun. Protective, work safety sunglasses are military tested and ANSI Compliant. Indoor/Outdoor Lens is ideal for indoors, outdoors, nighttime and hazy/cloudy conditions. Durable, nylon frames bend and flex without breaking. Nonslip, rubber nose piece and rubber temples keep the glasses secure and resist sweat. Frame width is 144mm.
Catalog No. 50-008-28417 Supplier Enterprise Technology Solutions Supplier No. EGO56080
Skullerz Loki Safety Glasses are designed to be worn as regular sunglasses or as safety goggles by removing the temples and attaching the elastic strap and removable foam gasket for a dust-beating seal (both included). Tough, scratch-resistant lens filters 99.9 percent of harmful UVA, UVB and UVC rays to keep your eyes safe in the sun. Protective, work safety sunglasses are military tested and ANSI Compliant. Indoor/Outdoor Lens is ideal for indoors, outdoors, nighttime and hazy/cloudy conditions. Durable, nylon frames bend and flex without breaking. Nonslip, rubber nose piece and rubber temples keep the glasses secure and resist sweat. Frame width is 144mm.