Learn More
Enterprise Technology Solutions Ergodyne Sunglasses/Goggles,Yellow Lens/Black Frame
SDP
Catalog No. 5000828083
Encompass_Preferred
Skullerz Loki Safety Glasses are designed to be worn as regular sunglasses or as safety goggles by removing the temples and attaching the elastic strap and removable foam gasket for a dust-beating seal (both included). Tough, scratch-resistant lens filters 99.9 percent of harmful UVA, UVB and UVC rays to keep your eyes safe in the sun. Protective, work safety sunglasses are military tested and ANSI Compliant. Ideal for indoors, outdoors, low light and hazy/cloudy conditions, this yellow lens increases brightness in dull conditions. Durable, nylon frames bend and flex without breaking. Nonslip, rubber nose piece and rubber temples keep the glasses secure and resist sweat. Frame width is 144mm.
Catalog No. 50-008-28083 Supplier Enterprise Technology Solutions Supplier No. EGO56050
Skullerz Loki Safety Glasses are designed to be worn as regular sunglasses or as safety goggles by removing the temples and attaching the elastic strap and removable foam gasket for a dust-beating seal (both included). Tough, scratch-resistant lens filters 99.9 percent of harmful UVA, UVB and UVC rays to keep your eyes safe in the sun. Protective, work safety sunglasses are military tested and ANSI Compliant. Ideal for indoors, outdoors, low light and hazy/cloudy conditions, this yellow lens increases brightness in dull conditions. Durable, nylon frames bend and flex without breaking. Nonslip, rubber nose piece and rubber temples keep the glasses secure and resist sweat. Frame width is 144mm.