Learn More
MSC Ergodyne Vest, Breakaway, ANSI Class 2, ID Holder, 1 ea
Catalog No. 5000767125
Encompass
This breakaway hi-vis safety vest is compliant with ANSI/ISEA 107-2020, Type R, Class 2 standards. It features 2 in reflective, solid tape for increased visibility in low-light conditions and is made from polyester mesh fabric (3.3 oz) for all-day comfort. The vest includes a hook and loop closure for adjustability and easy wear and removal, with breakaway shoulders and sides that detach if caught on equipment or machinery to prevent dangerous hang-ups.
  • ANSI/ISEA 107-2020, Type R, Class 2 compliant with 2 in reflective tape
  • Polyester mesh fabric (3.3 oz) for all-day comfort
  • Hook and loop closure for adjustability and easy wear
  • Breakaway shoulders and sides prevent dangerous snagging
  • Two pockets, badge holder, and one mic tab
  • Machine washable for easy care
Catalog No. 50-007-67125 Supplier MSC Supplier No. 39421953
This breakaway hi-vis safety vest is compliant with ANSI/ISEA 107-2020, Type R, Class 2 standards. It features 2 in reflective, solid tape for increased visibility in low-light conditions and is made from polyester mesh fabric (3.3 oz) for all-day comfort. The vest includes a hook and loop closure for adjustability and easy wear and removal, with breakaway shoulders and sides that detach if caught on equipment or machinery to prevent dangerous hang-ups.
  • ANSI/ISEA 107-2020, Type R, Class 2 compliant with 2 in reflective tape
  • Polyester mesh fabric (3.3 oz) for all-day comfort
  • Hook and loop closure for adjustability and easy wear
  • Breakaway shoulders and sides prevent dangerous snagging
  • Two pockets, badge holder, and one mic tab
  • Machine washable for easy care