Learn More
MSC Ergodyne XL Canvas Hoist Bucket, Swivel Carabiner, Top, 1 ea
Catalog No. 5000794077
Encompass
The Ergodyne Arsenal 5945T XL Canvas Hoist Bucket provides a reliable solution for safely hoisting, storing, and accessing tools at heights. Its durable 24 oz canvas construction, heavy-gauge nylon stitching, and synthetic leather reinforced bottom ensure long-lasting performance for larger tools.
  • Holds and protects up to 150 lb (68 kg) of tools at heights
  • Constructed from 24 oz canvas and synthetic leather reinforced base
  • Swiveling carabiner for tangle-free performance and drop prevention
  • Durable web handle with leather reinforcement for reliable hoisting
  • Molded top ring provides wide opening for easy tool access
  • One interior pocket for organization
  • Patented nickel-plated storm drain prevents rust and keeps tools dry
  • Includes Arsenal 5937 XL Hoist Bucket Top for tool security
Catalog No. 50-007-94077 Supplier MSC Supplier No. 39418710
The Ergodyne Arsenal 5945T XL Canvas Hoist Bucket provides a reliable solution for safely hoisting, storing, and accessing tools at heights. Its durable 24 oz canvas construction, heavy-gauge nylon stitching, and synthetic leather reinforced bottom ensure long-lasting performance for larger tools.
  • Holds and protects up to 150 lb (68 kg) of tools at heights
  • Constructed from 24 oz canvas and synthetic leather reinforced base
  • Swiveling carabiner for tangle-free performance and drop prevention
  • Durable web handle with leather reinforcement for reliable hoisting
  • Molded top ring provides wide opening for easy tool access
  • One interior pocket for organization
  • Patented nickel-plated storm drain prevents rust and keeps tools dry
  • Includes Arsenal 5937 XL Hoist Bucket Top for tool security