Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTL-Strefa DropSafe ErgoLance Micro Flow Lancet
SDP

Catalog No. 22192021
Change view
Click to view available options
Color:
Blue
Green
Pink
Needle Gauge:
21G
25G
30G
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Needle Gauge
22-192-021 Green 30G
22-192-022 Blue 25G
22-192-023 Pink 21G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-192-021 Supplier HTL-Strefa Supplier No. 8481
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sterile, single-use devices designed for the sampling of capillary blood from the fingertip of patients.

  • Special lancet shape distracts attention from needle stick
  • Concave housing for comfortable finger recess maximizes economy
  • No release button
  • Gentle pressure activation
  • Ribbed protective cap guarantees needle sterility and gives a secure grip for easy twist-off removal

Specifications

Product Type Safety Lancet
Quantity 100 ct. per box
Length (Metric) 1.5 mm
Color Green
For Use With (Application) Lancet for blood testing
Material Medical sharp
Needle Gauge 30G
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.