Learn More
Cayman Chemical ErgoloIdmesylates 100mg
SDP
Catalog No. 502933267
Encompass_Preferred
A mixture of alkaloids with diverse biological activities; blocks norepinephrine-induced reflex vasodilation and vasoconstriction in dogs when injected into the kidney at a dose of 0.15 mg; blocks norepinephrine-induced cAMP production in rat brain cortical homogenates ex vivo in a dose-dependent manner; acts as an erectogenic agent in rats
Catalog No. 50-293-3267 Supplier Cayman Chemical Supplier No. 24095100 MG
A mixture of alkaloids with diverse biological activities; blocks norepinephrine-induced reflex vasodilation and vasoconstriction in dogs when injected into the kidney at a dose of 0.15 mg; blocks norepinephrine-induced cAMP production in rat brain cortical homogenates ex vivo in a dose-dependent manner; acts as an erectogenic agent in rats