Learn More
Cayman Chemical Ergoloid (mesylates), Purity: ≥95%, CAS: 8067-24-1, 50 mg
SDP
Catalog No. 502933181
Encompass_Preferred
A mixture of alkaloids with diverse biological activities; blocks norepinephrine-induced reflex vasodilation and vasoconstriction in dogs when injected into the kidney at a dose of 0.15 mg; blocks norepinephrine-induced cAMP production in rat brain cortical homogenates ex vivo in a dose-dependent manner; acts as an erectogenic agent in rats
Catalog No. 50-293-3181 Supplier Cayman Chemical Supplier No. 2409550 MG
A mixture of alkaloids with diverse biological activities; blocks norepinephrine-induced reflex vasodilation and vasoconstriction in dogs when injected into the kidney at a dose of 0.15 mg; blocks norepinephrine-induced cAMP production in rat brain cortical homogenates ex vivo in a dose-dependent manner; acts as an erectogenic agent in rats