Learn More
Wesco Industrial Products DM-1100 Ergonomic Drum Handler
Catalog No. NC0776808 Shop All Wesco Industrial Products Products
Encompass_Preferred
Wesco Ergonomic Drum Handler #240150 1 Year Warranty, parts only. WESCO MADE hydraulic pump . Handles most 30, 55 or 85 gallon drums - Steel, Fiber and Poly. Works with most drums with locking rings. Lifts, transports, weighs and places drums on spill or standard pallets.U.S. Patented Gator Grip U.S. Patent No. 6,619,905 Spring-loaded jaws hold any 3/16" or higher drum rim. Operator does not need to touch drums to operate. Exclusive auto grip lock will not drop a drum if operated properly. (2) wheels and (2) swivel casters, 5" x 2" phenolic, and floor lock.
Catalog No. NC0776808 Supplier Wesco Industrial Products Supplier No. 240150
Add to Cart
Add to Cart
Request a Quote
Wesco Ergonomic Drum Handler #240150 1 Year Warranty, parts only. WESCO MADE hydraulic pump . Handles most 30, 55 or 85 gallon drums - Steel, Fiber and Poly. Works with most drums with locking rings. Lifts, transports, weighs and places drums on spill or standard pallets.U.S. Patented Gator Grip U.S. Patent No. 6,619,905 Spring-loaded jaws hold any 3/16" or higher drum rim. Operator does not need to touch drums to operate. Exclusive auto grip lock will not drop a drum if operated properly. (2) wheels and (2) swivel casters, 5" x 2" phenolic, and floor lock.