Learn More
Perkin Elmer US LLC Ergonomic Manual 11mm Hand Crimper
Catalog No. 502104600
Encompass_Preferred
PerkinElmer now offers a hand crimper and decapper developed with the user in mind. This robust and easy to use tool is ideal for consistent crimping and decapping of 11mm and 20mm caps - a must have for every lab!
Catalog No. 50-210-4600 Supplier Perkin Elmer US LLC Supplier No. N6621035
PerkinElmer now offers a hand crimper and decapper developed with the user in mind. This robust and easy to use tool is ideal for consistent crimping and decapping of 11mm and 20mm caps - a must have for every lab!