Learn More
Perkin Elmer US LLC Ergonomic Manual 20mm Hand Crimper
Catalog No. 502104596
Encompass_Preferred
We now offer a hand crimper and decapper developed with the user in mind. This robust and easy to use tool is ideal for consistant crimping and decapping of 11mm and 20mm caps - a must have for every lab!
Catalog No. 50-210-4596 Supplier Perkin Elmer US LLC Supplier No. N6621037
We now offer a hand crimper and decapper developed with the user in mind. This robust and easy to use tool is ideal for consistant crimping and decapping of 11mm and 20mm caps - a must have for every lab!