Learn More
World Precision Instrument Ergonomic Micro-Shear Flush Cutters, 12.7 cm, Black Blades
SDP
Catalog No. 502539295
Encompass
Size and shape of these cutters are optimized to work in high density areas. Rockwell hardness of 55-57 RC. Cutting capacity: 18 AWG (1.02 mm) soft wire.
Catalog No. 50-253-9295 Supplier World Precision Instrument Supplier No. 504749
Size and shape of these cutters are optimized to work in high density areas. Rockwell hardness of 55-57 RC. Cutting capacity: 18 AWG (1.02 mm) soft wire.