Learn More
World Precision Instrument Ergonomic Mini-Scissors, 13 cm
SDP
Catalog No. 502539294
Encompass
Innovative design of these cutters lessens operator hand strain. Cutting capacity: 20 AWG (0.8 mm) soft wire. 0.005’’ (0.127 mm) mylar, plastic and shielded cable.
Catalog No. 50-253-9294 Supplier World Precision Instrument Supplier No. 504750
Innovative design of these cutters lessens operator hand strain. Cutting capacity: 20 AWG (0.8 mm) soft wire. 0.005’’ (0.127 mm) mylar, plastic and shielded cable.