Learn More
MSC Vikan Ergonomic Shovel, Polypropylene, Green, 50 in
Catalog No. 502839331
Encompass
This ergonomic shovel is designed for moving large amounts of food waste or ingredients, promoting reduced strain with its lightweight and extended handle length. Made from durable polypropylene, it is suitable for various applications, including those requiring food contact compliance.
  • Ergonomic design helps reduce strain when lifting and shoveling.
  • Constructed with a durable polypropylene blade and anodized aluminum handle.
  • Features an extended handle length with a D-grip for comfortable use.
  • Made from FDA-compliant materials for food contact applications.
  • Can be integrated into a color-coding system for improved food safety.
  • Autoclavable and dishwasher safe for easy cleaning and sterilization.
  • Suitable for working in temperatures from -30°C to 80°C.
  • Complies with relevant EU and FDA regulations for food contact.
Catalog No. 50-283-9331 Supplier MSC Supplier No. 47170378
This ergonomic shovel is designed for moving large amounts of food waste or ingredients, promoting reduced strain with its lightweight and extended handle length. Made from durable polypropylene, it is suitable for various applications, including those requiring food contact compliance.
  • Ergonomic design helps reduce strain when lifting and shoveling.
  • Constructed with a durable polypropylene blade and anodized aluminum handle.
  • Features an extended handle length with a D-grip for comfortable use.
  • Made from FDA-compliant materials for food contact applications.
  • Can be integrated into a color-coding system for improved food safety.
  • Autoclavable and dishwasher safe for easy cleaning and sterilization.
  • Suitable for working in temperatures from -30°C to 80°C.
  • Complies with relevant EU and FDA regulations for food contact.