Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Ergonomic Training Solving The Puzzle Leaders Guide

Catalog No. 19103343 Shop All Brady Corporation Products
Change view
Click to view available options
Format:
Book
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19-103-343 Book
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-343 Supplier Brady™ Supplier No. 38095

Protect your workers from work-related musculoskeletal disorders with ergonomic training

Specifications

Format Book
Quantity Each
Languages English
For Use With (Application) Workplace Safety Training
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.