Learn More
Berkshire™ Ergonomic V Mop Foam Cover
Catalog No. 19070026 Shop All Berkshire Corporation Products
Change view
Click to view available options
Material:
Polyester Knit
Catalog No. Material
19-070-026 Polyester Knit
1 options
This item is not returnable. View return policy
Catalog No. 19-070-026 Supplier Berkshire™ Supplier No. FMC6LF12
Add to Cart
Add to Cart
This item is not returnable. View return policy
Abrasion resistant polyester knit clean room fabric laminated to absorbent polyurethane foam. Laser cut and cleanroom laundered to minimize fiber and particle release. 10 mm thickness provides cushioned surface for optimum conformance to uneven floors, walls and ceilings. High absorbency capacity.
Abrasion resistant polyester knit clean room fabric laminated to absorbent polyurethane foam. Laser cut and cleanroom laundered to minimize fiber and particle release. 10 mm thickness provides cushioned surface for optimum conformance to uneven floors, walls and ceilings. High absorbency capacity.
  • Abrasion resistant
  • Laser cut
  • High absorbency
Specifications
Product Type V Flat Mop Foam Cover
Sterility Non-sterile
Autoclavable Autoclavable
Color-coded No
Material Polyester Knit