Learn More
Berkshire™ Ergonomic V Mop Nonwoven Microfiber Cover
Catalog No. 19070025 Shop All Berkshire Corporation Products
Change view
Click to view available options
Material:
Polyester, Cellulose, Non-woven Rayon
Catalog No. Material
19-070-025 Polyester, Cellulose, Non-woven Rayon
1 options
Catalog No. 19-070-025 Supplier Berkshire™ Supplier No. FMC4NW20
Ergonomic Flat Mop Nonwoven Cover is made of polyester and cellulose with 100% rayon nonwoven inner layers to provide superior absorbency. Designed for the capture and removal of contaminants during the cleaning process. Each cover is individually wrapped to preserve cleanliness.
Ergonomic Flat Mop Nonwoven Cover is made of polyester and cellulose with 100% rayon nonwoven inner layers to provide superior absorbency. Designed for the capture and removal of contaminants during the cleaning process. Each cover is individually wrapped to preserve cleanliness.
  • 68 gsm polyester and cellulose outer cover
  • 100% rayon nonwoven inner layer
  • Individually wrapped to preserve cleanliness
  • Autoclavable
Specifications
Product Type V Flat Mop Nonwoven Cover
Sterility Non-sterile
Color-coded No
Material Polyester, Cellulose, Non-woven Rayon