Learn More
USA Scientific Inc ErgoOne 10-100ul 1 Chan
Catalog No. 502745348
Encompass
ErgoOne 10-100 uL single channel pipette, 0.1 uL increments
Catalog No. 50-274-5348 Supplier USA Scientific Inc Supplier No. 71001100
ErgoOne 10-100 uL single channel pipette, 0.1 uL increments