Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ ErgoPath Chair
SDP

Catalog No. 22445428 Shop All Mopec Products
Change view
Click to view available options
Material:
Chrome Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
22-445-428 Chrome Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-445-428 Supplier Mopec™ Supplier No. EC200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The ErgoPath Chair combines unparalleled, ergonomic lumbar support support with an elegant modern design.

  • Leather Seat (L x W): 17.5 x 18 in.
  • Leather Back Rest (H x W): 11 x 13 in.
  • Cylinder Heights: 16.25 to 21.25 in., 18.25 to 26 in., 21.25 to 31.5 in.
  • Height range: 16.25 to 31.5 in.
  • Weight Capacity: 350 to 400 lb.
  • Replaceable back pads

Specifications

Product Type Office Chair
Material Chrome Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.