Learn More
Foamtec ErgoSCRUB™ Scrubbing Tool
Catalog No. 19304775 Shop All Foamtec International Products
Change view
Click to view available options
Description:
Hard Handle
Soft Handle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
19-304-775 Hard Handle
19-120-2177 Soft Handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-304-775 Supplier Foamtec Supplier No. FT951
Add to Cart
Add to Cart
This item is not returnable. View return policy
Soft ergonomic scrubbing tool for implant tools, ceramic domes, Novellus heater blocks, and large vacuum chambers
  • Rounded, ergonomically designed scrubbing tool reduces hand stress and makes cleanroom scrubbing easier
  • Flexible soft handle for use on implant tools, heater blocks, vacuum chambers, and ceramic domes
  • Firmer hard handle provides uniform scrubbing action on flat surfaces, minimizng tool wear
  • Both accommodate a variety of diamond or silicone carbide ScrubDISK™ pads that attach with either pressure-sensitive adhesive or loop backing on reusable styles
Order Info
Available as a reusable or disposable tool. Contact your Fisher Scientific™ Customer Service Representative for details about ScrubDISK options.
Specifications
Product Type Scrubbing Tool
Description Hard Handle
Disposable Reusable
For Use With (Application) For cleaning critical surfaces
For Use With (Equipment) For implant tools, Ceramic domes, Novellus heater blocks and large vacuum chAmbers
Includes Loop backing