Learn More
Medchemexpress LLC HY-N0181 100mg Medchemexpress, Ergosterol CAS:57-87-4 Purity:>98%
SDP
Catalog No. 501933451
Encompass_Preferred
Medchemexpress, HY-N0181 100mg Ergosterol CAS:57-87-4 Ergosterol is the primary sterol found in fungi, with antioxidative, anti-proliferative, and anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3451 Supplier Medchemexpress LLC Supplier No. HYN0181100MG
Medchemexpress, HY-N0181 100mg Ergosterol CAS:57-87-4 Ergosterol is the primary sterol found in fungi, with antioxidative, anti-proliferative, and anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.