Learn More
TARGETMOL CHEMICALS INC Ergosterol Acetate 5MG
SDP
Catalog No. 502877146
Encompass_Preferred
Also available in 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ergosterol Acetate (Ergosta-5,7,22-trien-3-beta-yl acetate) is one of the acetate derivatives of Ergosterol from Monascus anka. Purity 98.44%
Catalog No. 50-287-7146 Supplier TARGETMOL CHEMICALS INC Supplier No. TN70715MG
Also available in 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ergosterol Acetate (Ergosta-5,7,22-trien-3-beta-yl acetate) is one of the acetate derivatives of Ergosterol from Monascus anka. Purity 98.44%