Learn More
Cayman Chemical Ergosterol Peroxide, Purity: ≥98%, CAS: 2061-64-5, 5 mg
SDP
Catalog No. 502988468
Encompass_Preferred
An oxidized sterol; active against the P. falciparum chloroquine-sensitive strain FCA 20/GHA (IC50 = 8.2 µM); induces apoptosis in LNCaP and DU145 prostate cancer cells from 6.25-50 µM; inhibits growth of SNU-1 gastric, SNU-354 hepatoma, and SNU-C4 colorectal cancer cells (IC50s = 18.7, 158.2, and 84.6 µM, respectively; inhibits LPS-induced NF-kB DNA-binding activity and production of TNF-α in RAW 264.7 cells at 60 µM; reduces ear edema in a mouse model of inflammation induced by TPA (ID50 = 0.2 mg/ear)
Catalog No. 50-298-8468 Supplier Cayman Chemical Supplier No. 308605 MG
An oxidized sterol; active against the P. falciparum chloroquine-sensitive strain FCA 20/GHA (IC50 = 8.2 µM); induces apoptosis in LNCaP and DU145 prostate cancer cells from 6.25-50 µM; inhibits growth of SNU-1 gastric, SNU-354 hepatoma, and SNU-C4 colorectal cancer cells (IC50s = 18.7, 158.2, and 84.6 µM, respectively; inhibits LPS-induced NF-kB DNA-binding activity and production of TNF-α in RAW 264.7 cells at 60 µM; reduces ear edema in a mouse model of inflammation induced by TPA (ID50 = 0.2 mg/ear)