Learn More
Ergosterol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 57-87-4 |
| Physical Form | Solid |
| Linear Formula | C28H44 O |
| MDL Number | MFCD00003623 |
| Melting Point | 156°C to 158°C |
| Synonym | Ergosterol; 3 beta-Hydroxy-5,7,22-ergostatriene; 5,7,22-Ergostatrien-3 beta-ol; Provitamin D2 |
| Type | Certified Reference Material |
| Quantity | 200 mg |
| Formula Weight | 396.65 |
| Grade | Certified Reference Material |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.