Learn More
TARGETMOL CHEMICALS INC ERGOTHIONEINE 25MG
SDP
Catalog No. 502579095
Encompass_Preferred
Also available in 10 mg 50 mg and bulk. Please contact Fisher for quotes. Ergothioneine (L-(+)-Ergothioneine) is synthesized by certain bacteria and fungi. It is generally considered an antioxidant. purity: 98%
Catalog No. 50-257-9095 Supplier TARGETMOL CHEMICALS INC Supplier No. T819125MG
Also available in 10 mg 50 mg and bulk. Please contact Fisher for quotes. Ergothioneine (L-(+)-Ergothioneine) is synthesized by certain bacteria and fungi. It is generally considered an antioxidant. purity: 98%