Learn More
Medchemexpress LLC Ergothioneine-d9 | 497-30-3 | 238.35 | 1 MG
SDP
Catalog No. 5000415429
Encompass_Preferred
Ergothioneine-d9 is the deuterium labeled Ergothioneine. Ergothioneine, an imidazole-2-thione derivative of histidine betaine, is synthesized by certain bacteria and fungi. Ergothioneine is generally considered an antioxidant.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-15429 Supplier Medchemexpress LLC Supplier No. HYN1914S11MG
Ergothioneine-d9 is the deuterium labeled Ergothioneine. Ergothioneine, an imidazole-2-thione derivative of histidine betaine, is synthesized by certain bacteria and fungi. Ergothioneine is generally considered an antioxidant.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS