Learn More
Aviva Systems Biology Corp ERH Antibody - middle region (ARP48260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERH. It was validated on Western Blot. . Alternate names: DROER, FLJ27340ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-354