Learn More
Medchemexpress LLC HY-N0517 5mg Medchemexpress, Erianin CAS:95041-90-0 Purity:>98%
SDP
Catalog No. 501933602
Encompass_Preferred
Medchemexpress, HY-N0517 5mg Erianin CAS:95041-90-0 Erianin, often used as an antipyretic and analgesic agent, could inhibit IDO-induced tumor angiogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3602 Supplier Medchemexpress LLC Supplier No. HYN05175MG
Medchemexpress, HY-N0517 5mg Erianin CAS:95041-90-0 Erianin, often used as an antipyretic and analgesic agent, could inhibit IDO-induced tumor angiogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.