Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eribulin | 253128-41-5 | MFCD23160037 | 729.90 g/mol | C40H59NO11 | 500 UG
SDP

Catalog No. 5000203684
Encompass_Preferred

Eribulin is a microtubule-targeting research compound used to study metastatic breast cancer and apoptosis mechanisms. It is supplied as a dry powder soluble in DMSO, and requires cold, light-protected storage under inert atmosphere.

  • Microtubule inhibitor used in cell cycle and apoptosis research.
  • Molecular formula C40H59NO11 and molecular weight 729.90 g/mol.
  • Soluble in DMSO at high concentrations (200 mg/mL).
  • Requires storage at low temperatures and protection from light.
  • Suitable for use as an ADC payload or cytotoxic reference compound in research.

Catalog No. 50-002-03684 Supplier Medchemexpress LLC Supplier No. HY13442500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eribulin is a microtubule-targeting research compound used to study metastatic breast cancer and apoptosis mechanisms. It is supplied as a dry powder soluble in DMSO, and requires cold, light-protected storage under inert atmosphere.

  • Microtubule inhibitor used in cell cycle and apoptosis research.
  • Molecular formula C40H59NO11 and molecular weight 729.90 g/mol.
  • Soluble in DMSO at high concentrations (200 mg/mL).
  • Requires storage at low temperatures and protection from light.
  • Suitable for use as an ADC payload or cytotoxic reference compound in research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.