Learn More
Apexbio Technology LLC ERIBULIN-MESYLATE-10MG
SDP
Catalog No. 5000567988
Encompass_Preferred
Eribulin mesylate (CAS No 441045-17-6) is a synthetic analog of halichondrin B derived from a marine sponge primarily used in cancer research as a microtubule dynamics inhibitor Its mechanism of action involves the inhibition of microtubule growth without affecting the shortening phase which leads to cell cycle arrest and apoptosis Eribulin mesylate exhibits potent in vitro activity generally in the nanomolar range making it highly effective in various cancer cell lines It is commonly employed in cell models and animal studies to evaluate its potential therapeutic effects and mechanisms Additionally it is valuable in drug screening for its unique action on the mitotic spindle Experimental concentrations typically depend on the specific research design allowing flexibility based on the requirements of particular studies
Catalog No. 50-005-67988 Supplier Apexbio Technology LLC Supplier No. B338610
Eribulin mesylate (CAS No 441045-17-6) is a synthetic analog of halichondrin B derived from a marine sponge primarily used in cancer research as a microtubule dynamics inhibitor Its mechanism of action involves the inhibition of microtubule growth without affecting the shortening phase which leads to cell cycle arrest and apoptosis Eribulin mesylate exhibits potent in vitro activity generally in the nanomolar range making it highly effective in various cancer cell lines It is commonly employed in cell models and animal studies to evaluate its potential therapeutic effects and mechanisms Additionally it is valuable in drug screening for its unique action on the mitotic spindle Experimental concentrations typically depend on the specific research design allowing flexibility based on the requirements of particular studies