Learn More
TARGETMOL CHEMICALS INC Eribulin mesylate 1MG
SDP
Catalog No. 502875953
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Eribulin mesylate (E7389 mesylate) (E7389 mesylate) inhibits the proliferation of cancer cells by binding microtubule proteins and microtubules. Eribulin mesylate (E7389 mesylate) is a microtubule targeting agent that is used in the treatment of metastatic breast cancer. Purity 97.02%
Catalog No. 50-287-5953 Supplier TARGETMOL CHEMICALS INC Supplier No. T136871MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Eribulin mesylate (E7389 mesylate) (E7389 mesylate) inhibits the proliferation of cancer cells by binding microtubule proteins and microtubules. Eribulin mesylate (E7389 mesylate) is a microtubule targeting agent that is used in the treatment of metastatic breast cancer. Purity 97.02%