Learn More
Medchemexpress LLC Eribulin mesylate | 441045-17-6 | 99.3% | 826.00 | 10 MG
SDP
Catalog No. 5000365601
Encompass_Preferred
Eribulin mesylate is a microtubule targeting agent designed for metastatic breast cancer research. It effectively inhibits the proliferation of cancer cells by binding to microtubule proteins and microtubules, demonstrating significant activity both in vitro and in vivo.
  • Inhibits cell proliferation in various cancer cell lines.
  • Induces early apoptosis and G2/M arrest in LM8 cells.
  • Suppresses cell migration at low concentrations.
  • Reduces primary tumor growth and lung metastasis in osteosarcoma models.
  • Suppresses circulating tumor cells.
Catalog No. 50-003-65601 Supplier Medchemexpress LLC Supplier No. 10MGHY13442A
Eribulin mesylate is a microtubule targeting agent designed for metastatic breast cancer research. It effectively inhibits the proliferation of cancer cells by binding to microtubule proteins and microtubules, demonstrating significant activity both in vitro and in vivo.
  • Inhibits cell proliferation in various cancer cell lines.
  • Induces early apoptosis and G2/M arrest in LM8 cells.
  • Suppresses cell migration at low concentrations.
  • Reduces primary tumor growth and lung metastasis in osteosarcoma models.
  • Suppresses circulating tumor cells.